Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydro-2H-pyran-3-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR70150-1G OR70150-5G
76587-520EA 85 USD InStock
76587-520 76587-518
Tetrahydro-2H-pyran-3-carboxylic acid
Tetrahydro-2H-pyran-3-carboxylic acid


Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 257…258 °C/750mmHg
Density: 1.685 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD04114349
CAS Number: 873397-34-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty