Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (S)-tert-Butyl pyrrolidin-3-ylcarbamate(3S)-(-)-3-(Boc-amino)pyrrolidine

OR4636-5G OR4636-25G OR4636-100G
76585-018EA 268 USD InStock
76585-018 76585-014 76585-016
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine


Formula: C₉H₁₈N₂O₂
MW: 186.26 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00143194
CAS Number: 122536-76-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty