Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) 99%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: LAPPhenyl(2,4,6-trimethylbenzoyl)phosphinic acid lithium saltLithium PTMB phosphinate

OR48106-1G
76585-730EA 126 USD InStock
76585-730
Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) 99%
Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP)


Formula: C₁₆H₁₆LiO₃P
MW: 294.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29472537
CAS Number: 85073-19-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty