Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-4'-chlorobiphenyl

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4'-Chloro-[1,1'-biphenyl]-2-amine

OR7338-10MG
76587-900EA 13 USD InStock
76587-900
2-Amino-4'-chlorobiphenyl
2-Amino-4'-chlorobiphenyl


Formula: C₁₂H₁₀ClN
MW: 203.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424659
CAS Number: 1204-44-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty