Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromo-3-methylpyridine-2-carboxaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 5-Bromo-3-methylpicolinaldehyde5-Bromo-3-methyl-2-pyridinecarbaldehyde5-Bromo-2-formyl-3-methylpyridine

OR51763-1G OR51763-250MG
76586-700EA 28 USD InStock
76586-700 76586-702
5-Bromo-3-methylpyridine-2-carboxaldehyde 97%
5-Bromo-3-methylpyridine-2-carboxaldehyde


Formula: C₇H₆BrNO
MW: 200.033 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07776420
CAS Number: 376587-53-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty