Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Acetoxybenzyl alcohol

Supplier: APOLLO SCIENTIFIC
Warning

OR51991-1G OR51991-5G
76586-918EA 231 USD InStock
76586-918 76586-916
4-Acetoxybenzyl alcohol
4-Acetoxybenzyl alcohol


Formula: C₉H₁₀O₃
MW: 166.174 g/mol
Melting Pt: 37…42 °C
Density: 1.169 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02752162
CAS Number: 6309-46-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty