Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyrazine 95+%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-[3-(1,4-Diazin-2-yl)-1,2,4-oxadiazol-5-yl]piperidine5-(Piperidin-4-yl)-3-(pyrazin-2-yl)-1,2,4-oxadiazole

OR7807-250MG
76588-284EA 23 USD InStock
76588-284
2-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyrazine 95+%
2-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyrazine


Formula: C₁₁H₁₃N₅O
MW: 231.254 g/mol
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD03426077
CAS Number: 849925-00-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty