Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Falcon® Feeder Tray, 24-Well, Corning
Supplier: Corning
Falcon®
351186 351186
BD351186CS 118.41 USD InStock
CA-BD351186 BD351186
Falcon® Feeder Tray, 24-Well, Corning
Trays/Dishes/Pots Laboratory Trays/Dishes
Large volume media feeder tray reduces the need for frequent media changes. Feeder tray allows all 24 wells to be bathed in the same medium for better uniformity across the plate.
For use with BD BioCoat* HTS Caco-2 assay systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty