Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc 99+%, mossy

Supplier: APOLLO SCIENTIFIC
Warning

IN3874-5KG IN3874-1KG
76615-114EA 123 USD InStock
76615-114 76615-116
Zinc 99+%, mossy
Zinc


Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908 °C
Melting Pt: 419 °C
Density: 7.133 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty