Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Dimethoxytetrahydrofuran 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR43635-1KG OR43635-500G OR43635-100G
76611-850EA 43 USD InStock
76611-850 76611-852 76611-854
2,5-Dimethoxytetrahydrofuran 98%
2,5-Dimethoxytetrahydrofuran


Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 145…148 °C
Melting Pt: –45 °C
Density: 1.021 g/cm³ (25 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00005359
CAS Number: 696-59-3
EINECS: 211-797-1
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty