Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(-)-Epichlorhydrin 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: (R)-(-)-Glycerol epichlorhydrin(R)-(-)-3-Chloro-1,2-epoxypropane(R)-(-)-1-Chloro-2,3-epoxypropane

OR28970-500G OR28970-100G OR28970-25G
76611-522EA 63 USD InStock
76611-522 76611-524 76611-526
(R)-(-)-Epichlorhydrin 99%
(R)-(-)-Epichlorhydrin


Formula: ClCH₂CH(O)CH₂
MW: 92.52 g/mol
Boiling Pt: 114…117 °C
Density: 1.18 g/cm³ (25 °C)
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00077759
CAS Number: 51594-55-9
EINECS: 424-280-2
UN: 2023
ADR: 6.1+3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty