Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9,10-Dibromoanthracene

Supplier: APOLLO SCIENTIFIC
Warning

OR30417-25G OR30417-100G
76616-350EA 38 USD InStock
76616-350 76616-348
9,10-Dibromoanthracene
9,10-Dibromoanthracene


Formula: C₁₄H₈Br₂
MW: 336.03 g/mol
Melting Pt: 222…224 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00001244
CAS Number: 523-27-3
EINECS: 208-342-4
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty