Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Dibromoaniline 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR2212-5G OR2212-25G OR2212-100G
76616-044EA 53 USD InStock
76616-044 76616-042 76616-046
2,6-Dibromoaniline 98%
2,6-Dibromoaniline


Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Boiling Pt: 262…264 °C
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00007638
CAS Number: 608-30-0
EINECS: 210-158-4
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty