Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-4-nitrobenzene 98+%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 4-Chloronitrobenzene

OR5413-250G OR5413-1KG OR5413-5KG
76612-062EA 58 USD InStock
76612-062 76612-060 76612-064
1-Chloro-4-nitrobenzene 98+%
1-Chloro-4-nitrobenzene


Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 242 °C
Melting Pt: 80…83 °C
Density: 1.298 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007285
CAS Number: 100-00-5
UN: 1578
ADR: 6.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty