Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Bromo-2-chlorotoulene 97%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2-Chlorobenzyl bromide1-(Bromomethyl)-2-chlorobenzene

OR5033-100G OR5033-25G
76621-494EA 99 USD InStock
76621-494 76621-496
α-Bromo-2-chlorotoulene 97%
α-Bromo-2-chlorotoulene


Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 103…104 °C/10mmHg
Density: 1.583 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00000566
CAS Number: 611-17-6
EINECS: 210-257-2
UN: 3265
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty