Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Quinolinol

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 8-Hydroxyquinoline

OR2837-100G OR2837-500G OR2837-1KG
76616-230EA 45 USD InStock
76616-230 76616-228 76616-226
8-Quinolinol
8-Quinolinol


Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 267 °C/252mmHg
Melting Pt: 70…76 °C
Density: 1.03 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006807
CAS Number: 148-24-3
EINECS: 205-711-1
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty