Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorocyclopentane

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Cyclopentyl chloride

OR10099-500G OR10099-100G
76610-906EA 120 USD InStock
76610-906 76610-904
Chlorocyclopentane
Chlorocyclopentane


Formula: C₅H₉Cl
MW: 104.58 g/mol
Boiling Pt: 113…114 °C
Melting Pt: –94 °C
Density: 1.005 g/cm³ (25 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00001360
CAS Number: 930-28-9
EINECS: 213-212-5
UN: 1993
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty