Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopropylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR18344-250G OR18344-100G
76611-268EA 42 USD InStock
76611-268 76611-270
Cyclopropylamine 98%
Cyclopropylamine


Formula: C₃H₇N
MW: 57.1 g/mol
Boiling Pt: 49…50 °C
Melting Pt: –10 °C
Density: 0.824 g/cm³ (25 °C)
Flash Pt: –25 °C
Storage Temperature: Ambient
MDL Number: MFCD00001301
CAS Number: 765-30-0
UN: 2733
ADR: 3+8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty