Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antimony(III) chloride 99,9%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Antimony trichloride

IN1108-25G IN1108-100G
76610-770EA 114 USD InStock
76610-770 76610-768
Antimony(III) chloride 99,9%
Antimony (III) chloride


Formula: SbCl₃
MW: 228.12 g/mol
Boiling Pt: 283 °C
Melting Pt: 73.4 °C
Density: 3.14 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty