Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Chlorosuccinimide 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 1-Chloropyrrolidine-2,5-dione

OR0856-500G OR0856-1KG
76615-266EA 56 USD InStock
76615-266 76615-268
N-Chlorosuccinimide 99%
N-Chlorosuccinimide


Formula: C₄H₄ClNO₂
MW: 133.53 g/mol
Melting Pt: 148…150 °C
Density: 1.65 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005511
CAS Number: 128-09-6
EINECS: 204-878-8
UN: 3261
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty