Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(I) iodide 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Cuprous iodide

IN1559-1KG IN1559-500G IN1559-100G
76615-080EA 126 USD InStock
76615-080 76615-082 76615-078
Copper(I) iodide 98%
Copper (I) iodide


Formula: CuI
MW: 190.45 g/mol
Boiling Pt: 1290 °C
Melting Pt: 605 °C
Density: 5.62 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010978
CAS Number: 7681-65-4
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty