Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2'-Bipyridyl 99+%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2,2'-Bipyridine2,2'-Dipyridylα,α′-Dipyridyl

OR28749-250G OR28749-100G OR28749-25G
76616-252EA 13 USD InStock
76616-252 76616-250 76616-248
2,2'-Bipyridyl 99+%
2,2'-Bipyridyl


Formula: C₁₀H₈N₂
MW: 156.19 g/mol
Boiling Pt: 273 °C
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00006212
CAS Number: 366-18-7
EINECS: 206-674-4
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty