Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromovaleryl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 5-Bromovalerylchloride5-Bromopentanoyl chloride

OR18308-5G OR18308-25G
76620-718EA 57 USD InStock
76620-718 76620-716
5-Bromovaleryl chloride 95%
5-Bromovaleryl chloride


Formula: BrH₂C(CH₂)₃COOCl
MW: 199.47 g/mol
Boiling Pt: 116…118 °C/33mmHg
Density: 1.49 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00013660
CAS Number: 4509-90-4
UN: 3265
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty