Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(1R,2R)-(-)-1,2-Diaminocyclohexane 97%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: (1R,2R)-Cyclohexane-1,2-diamine(R,R)-DACH

OR307156-5G OR307156-25G OR307156-1G
76616-384EA 19.57 USD InStock
76616-384 76616-388 76616-386
(1R,2R)-(-)-1,2-Diaminocyclohexane 97%
(1R,2R)-(-)-1,2-Diaminocyclohexane
Chiral intermediate for pharmaceuticals and agrochemicals.


Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 104…110 °C/40mmHg
Melting Pt: 40…43 °C
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00062985
CAS Number: 20439-47-8
UN: 3259
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty