Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium hydroxide octahydrate 99.99%

Supplier: APOLLO SCIENTIFIC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger

IN1198-25G IN1198-100G
76620-622EA 57 USD InStock
76620-622 76620-620
Barium hydroxide octahydrate 99.99%
Barium hydroxide octahydrate


Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Melting Pt: 78 °C
Density: 2.18 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
UN: 3262
ADR: 8,II
Order Now

 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty