Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Hydroxythiophenol

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: m-Hydroxythiophenol3-Hydroxybenzene-1-thiol3-Hydroxybenzenethiol3-Mercaptophenol3-Hydroxyphenyl mercaptan3-Hydroxythiophenol1-Hydroxy-3-mercaptobenzene

OR0777-5G OR0777-25G OR0777-100G
76624-662EA 404 USD InStock
76624-662 76624-664 76624-666
3-Hydroxythiophenol
3-Hydroxythiophenol


Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 242…243 °C
Melting Pt: 16…17 °C
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01317829
CAS Number: 40248-84-8
UN: 3334
ADR: 9,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty