Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylmorpholine

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: N-MethylmorpholineNMM

OR16938-500ML OR16938-100ML
76611-220EA 63 USD InStock
76611-220 76611-218
4-Methylmorpholine
4-Methylmorpholine


Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 115…116 °C
Melting Pt: –66 °C
Density: 0.92 g/cm³ (25 °C)
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00006175
CAS Number: 109-02-4
EINECS: 203-640-0
UN: 2535
ADR: 3+8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty