Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(-)-1-Phenylethanol 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: (S)-(-)-sec-Phenethyl alcohol(S)-1-Phenylethanol(S)-(-)-α-Methylbenzyl alcohol

OR315315-100G OR315315-25G
76616-440EA 21 USD InStock
76616-440 76616-438
(S)-(-)-1-Phenylethanol 99%
(S)-(-)-1-Phenylethanol


Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 98 °C/20.25mmHg
Density: 0.996 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00064264
CAS Number: 1445-91-6
UN: 2937
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty