Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dimethylphenyl isocyanate

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 3,5-Dimethylphenylisocyanate3,5-Xylyl isocyanate

OR0415-25G OR0415-100G OR0415-5G
76610-852EA 19 USD InStock
76610-852 76610-850 76610-848
3,5-Dimethylphenyl isocyanate
3,5-Dimethylphenyl isocyanate
  • May form small amount of sediment.


Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 205 °C
Density: 1.045 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013868
CAS Number: 54132-75-1
EINECS: 258-987-0
UN: 2206
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty