Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dichlorobenzylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR30090-1G OR30090-25G OR30090-5G
76611-570EA 132 USD InStock
76611-570 76611-572 76611-568
3,5-Dichlorobenzylamine 98%
3,5-Dichlorobenzylamine


Formula: C₇H₇Cl₂N
MW: 176.04 g/mol
Boiling Pt: 141…144 °C/15mmHg
Density: 1.32 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00052681
CAS Number: 39989-43-0
UN: 2735
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty