Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Hexynoic acid 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR30593-25G OR30593-1G OR30593-5G
76621-054EA 227 USD InStock
76621-054 76621-056 76621-052
5-Hexynoic acid 99%
5-Hexynoic acid


Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 224…225 °C
Density: 1.016 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00066346
CAS Number: 53293-00-8
UN: 3265
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty