Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) bromide, Reagent Grade

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Cuprous bromideCupric bromide

IN1540-100G
76615-076EA 28 USD InStock
76615-076
Copper(II) bromide, Reagent Grade
Copper (II) bromide


Formula: Br₂Cu
MW: 223.35 g/mol
Boiling Pt: 900 °C
Melting Pt: 498 °C
Density: 4.71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010970
CAS Number: 7789-45-9
EINECS: 232-167-2
UN: 3260
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty