Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Dimethylamino)benzoyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR11167-5G OR11167-25G
76615-420EA 161.71 USD InStock
76615-420 76615-422
4-(Dimethylamino)benzoyl chloride
4-(Dimethylamino)benzoyl chloride


Formula: C₉H₁₀ClNO
MW: 183.64 g/mol
Boiling Pt: 180 °C/15mmHg
Melting Pt: 142…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00236039
CAS Number: 4755-50-4
UN: 3261
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty