Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Phenoxybenzyl alcohol

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (3-Phenoxyphenyl)methanol3-(Hydroxymethyl)diphenyl ether

OR18354-100G OR18354-500G OR18354-25G
76615-920EA 90 USD InStock
76615-920 76615-916 76615-918
3-Phenoxybenzyl alcohol
3-Phenoxybenzyl alcohol


Formula: C₁₃H₁₂O₂
MW: 200.24 g/mol
Boiling Pt: 135…140 °C/0.1mmHg
Melting Pt: 44381 °C
Density: 1.149 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004636
CAS Number: 13826-35-2
EINECS: 237-525-1
UN: 3082
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty