Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl glyoxime 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Diacetyl dioxime2,3-Butanedione dioximeDiacetyl glyoxime

OR14325-100G OR14325-500G OR14325-2.5KG
76615-710EA 66 USD InStock
76615-710 76615-706 76615-708
Dimethyl glyoxime 99%
Dimethyl glyoxime


Formula: CH₃C(=NOH)C(=NOH)CH₃
MW: 116.12 g/mol
Melting Pt: 240…241 °C
Storage Temperature: Ambient
MDL Number: MFCD00002117
CAS Number: 95-45-4
EINECS: 202-420-1
UN: 2926
ADR: 4.1+6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty