Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopropyl methyl ketone

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 1-Cyclopropylethanone

OR10100-500G OR10100-25G OR10100-100G
76610-908EA 57 USD InStock
76610-908 76610-912 76610-910
Cyclopropyl methyl ketone
Cyclopropyl methyl ketone


Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 113…114 °C
Melting Pt: < –70 °C
Density: 0.898 g/cm³ (25 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00001297
CAS Number: 765-43-5
EINECS: 212-146-4
UN: 1992
ADR: 3+6.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty