Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Methyltetrahydrofuran 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR42160-500ML OR42160-1L OR42160-5X1L
76611-840EA 40 USD InStock
76611-840 76611-842 76611-838
(±)-2-Methyltetrahydrofuran 99%
(±)-2-Methyltetrahydrofuran


Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 78…82 °C
Melting Pt: –136 °C
Density: 0.856 g/cm³ (25 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00005367
CAS Number: 96-47-9
EINECS: 202-507-4
UN: 2536
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty