Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(4-methoxyphenyl) disulfide 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 1,2-Bis(4-methoxyphenyl)disulfane1,2-Bis(4-methoxyphenyl)disulphane

OR11235-1G OR11235-5G OR11235-25G
76615-442EA 186 USD InStock
76615-442 76615-444 76615-440
Bis(4-methoxyphenyl) disulfide 99%
Bis(4-methoxyphenyl) disulphide


Formula: C₁₄H₁₄O₂S₂
MW: 278.4 g/mol
Boiling Pt: 200 °C/3mmHg
Melting Pt: 40…45 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00041358
CAS Number: 5335-87-5
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty