Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorostyrene

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-Chloro-2-vinylbenzene

OR0031-25G OR0031-100G
76610-812EA 212 USD InStock
76610-812 76610-814
2-Chlorostyrene
2-Chlorostyrene


Formula: C₈H₇Cl
MW: 138.6 g/mol
Boiling Pt: 58…60 °C/7mmHg
Melting Pt: –63 °C
Density: 1.08 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000567
CAS Number: 2039-87-4
EINECS: 218-026-8
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty