Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diisopropylcarbodiimide 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: N,N'-methanetetraylbis(1-methylethylamine)1,3-DiisopropylcarbodiimideN,N-DiisopropylcarbodiimideN,N'-DiisopropylcarbodiimideN.N’-DiisopropylcarbodiimideDIC

OR18304-100G OR18304-25G
76615-890EA 70 USD InStock
76615-890 76615-892
Diisopropylcarbodiimide 99%
Diisopropylcarbodiimide


Formula: C₇H₁₄N₂
MW: 126.2 g/mol
Boiling Pt: 145…148 °C
Density: 0.815 g/cm³ (25 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00065689
CAS Number: 693-13-0
EINECS: 211-743-7
UN: 2929
ADR: 6.1+3,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty