Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-2-thiophenesulfonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 5-Chlorothiophene-2-sulfonyl chloride5-Chlorothiophene-2-sulphonyl chloride

OR5379-5G OR5379-25G
76621-600EA 63 USD InStock
76621-600 76621-602
5-Chloro-2-thiophenesulfonyl chloride 95%
5-Chloro-2-thiophenesulphonyl chloride


Formula: C₄H₂Cl₂O₂S₂
MW: 217.1 g/mol
Boiling Pt: 112…117 °C/15mmHg
Melting Pt: 26…28 °C
Density: 1.623 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00051667
CAS Number: 2766-74-7
UN: 3261
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty