Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(2-Aminoethyl)-4-methylpiperazine 95%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2-(4-Methyl-piperazin-1-yl)-ethylamine2-(4-Methylpiperazin-1-yl)ethanamine

OR0800-5G OR0800-1G
76610-868EA 48 USD InStock
76610-868 76610-866
1-(2-Aminoethyl)-4-methylpiperazine 95%
1-(2-Aminoethyl)-4-methylpiperazine


Formula: C₇H₁₇N₃
MW: 143.23 g/mol
Boiling Pt: 112 °C/30mmHg
Storage Temperature: Ambient
MDL Number: MFCD03701701
CAS Number: 934-98-5
EINECS: 213-296-3
UN: 2735
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty