Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(cyclopentadienyl)titanium dichloride

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Titanocene dichlorideBis(cyclopentadienyl)titanium(IV) dichlorideTitanocenedichloride

OR17913-10G OR17913-50G OR17913-250G
76615-866EA 13 USD InStock
76615-866 76615-868 76615-870
Bis(cyclopentadienyl)titanium dichloride
Bis(cyclopentadienyl)titanium dichloride


Formula: C₁₀H₁₀Cl₂Ti
MW: 248.97 g/mol
Melting Pt: 260…280 °C (dec.)
Density: 1.6 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003723
CAS Number: 1271-19-8
EINECS: 215-035-9
UN: 3261
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty