Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenyliodonium chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR451320-5G OR451320-25G
76616-770EA 19 USD InStock
76616-770 76616-768
Diphenyliodonium chloride
Diphenyliodonium chloride


Formula: C₁₂H₁₀ClI
MW: 316.57 g/mol
Melting Pt: 233…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00011909
CAS Number: 1483-72-3
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty