Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: (S)-1,2,3,4-Tetrahydronaphthalen-1-amine

OR40592-25G OR40592-5G OR40592-1G
76611-820EA 13.39 USD InStock
76611-820 76611-822 76611-824
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine


Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Boiling Pt: 250 °C
Density: 1.01 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00671630
CAS Number: 23357-52-0
UN: 2735
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty