Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-3-amino-1H-pyrazole-4-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Amino-4-carbethoxypyrazoleEthyl 3-amino-1H-pyrazole-4-carboxylate

OR13363-25G OR13363-5G
76615-640EA 13.39 USD InStock
76615-640 76615-638
Ethyl-3-amino-1H-pyrazole-4-carboxylate
Ethyl 3-amino-1H-pyrazole-4-carboxylate


Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00005238
CAS Number: 6994-25-8
EINECS: 230-262-3
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty