Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-ethylenediamine 97+%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: t-boc-N-EDAtert-Butyl N-(2-Aminoethyl)carbamateN-(tert-Butoxycarbonyl)-1,2-ethylenediaminetert-Butyl (2-aminoethyl)carbamateN-(tert-Butoxycarbonyl)-1,2-diaminoethaneN-BOC-1,2-diaminoethane

OR1533-25G OR1533-100G OR1533-5G
76611-152EA 51 USD InStock
76611-152 76611-154 76611-156
N-Boc-ethylenediamine 97+%
N-Boc-ethylenediamine


Formula: C₇H₁₆N₂O₂
MW: 160.22 g/mol
Boiling Pt: 72…80 °C/0.1mmHg
Density: 1.012 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
UN: 2735
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty