Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methoxy-4-methylbenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR11330-100G OR11330-5G OR11330-25G
76624-718EA 326 USD InStock
76624-718 76624-720 76624-722
3-Methoxy-4-methylbenzonitrile 98%
2-Methoxy-5-methylbenzonitrile


Formula: C₉H₉NO
MW: 147.1739 g/mol
Melting Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD03428590
CAS Number: 3556-60-3
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty