Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3,5-Triethynylbenzene 98+%

Supplier: APOLLO SCIENTIFIC
Danger

OR13102-5G OR13102-1G
76611-076EA 16 USD InStock
76611-076 76611-078
1,3,5-Triethynylbenzene 98+%
1,3,5-Triethynylbenzene


Formula: C₁₂H₆
MW: 150.18 g/mol
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00068621
CAS Number: 7567-63-7
UN: 1325
ADR: 4.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty