Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-3,4-dimethylbenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR5275-25G OR5275-5G
76591-356EA 13 USD InStock
76591-356 76591-358
2-Amino-3,4-dimethylbenzoic acid
2-Amino-3,4-dimethylbenzoic acid


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00130041
CAS Number: 50419-58-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty